BACEA-4465

4028495.0001 4028495.0005
A-4465.0005BAEA 359.08 USD
A-4465.0005BA A-4465.0001BA
BACEA-4465
Sequence: Boc-cyclohexyl-Gly-OH

Bachem offers derivatives of α-aminoglycine, cyclohexylglycine, cyclopentylglycine, and the PNA building block N-β-aminoethylglycine. For t-butylglycine derivatives see t-Leucine.
Chg derivatives.

Synonym(s): Boc-Chg-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR