Boc-D-Asn(Xan)-OH

4028747.0005
A-4030.0005BAEA 482.28 USD
A-4030.0005BA
Boc-D-Asn(Xan)-OH
Boc-D-Asn(Xan)-OH

Sequence: Boc-D-Asn(Xan)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₂₂H₂₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00076910
CAS Number: 200192-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR