Boc-D-Tyr(2,6-CL2-BZL)-OH

Synonyms: Boc-D-Tyr(2,6-dichloro-Bzl)-OH

4030419.0005
A-4025.0005BAEA 308.93 USD
A-4025.0005BA
Boc-D-Tyr(2,6-CL2-BZL)-OH
Boc-D-Tyr(2,6-CL2-BZL)-OH

Sequence: Boc-D-Tyr(2,6-dichloro-Bzl)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₁H₂₃Cl₂NO₅
MW: 440.317 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037107
CAS Number: 69541-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR