Boc-N-methyl-D-Valine

Synonyms: N-Boc-N-methyl-D-valine(R)-2-((tert-Butoxycarbonyl)(methyl)amino)-3-methylbutanoic acidBoc-N-Me-D-Val-OH

4028192.0001
A-3980.0001BAEA 121.26 USD
A-3980.0001BA
Boc-N-methyl-D-Valine
Boc-N-methyl-D-Valine

Sequence: Boc-N-Me-D-Val-OH


Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
MeVal, αMeVal.

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076996
CAS Number: 89536-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR