Boc-D-pentafluorophenylalanine

Synonyms: Boc-pentafluoro-D-Phe-OH

4030417.0001
A-3960.0001BAEA 187.62 USD
A-3960.0001BA
Boc-D-pentafluorophenylalanine
Boc-D-pentafluorophenylalanine

Sequence: Boc-pentafluoro-D-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₁₄H₁₄F₅NO₄
MW: 355.261 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151892
CAS Number: 136207-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR