N-Boc-S-benzyl-D-cysteine

Synonyms: Boc-D-Cys(Bzl)-OH

4030402.0005
A-3905.0005BAEA 261.79 USD
A-3905.0005BA
N-Boc-S-benzyl-D-cysteine
N-Boc-S-benzyl-D-cysteine

Sequence: Boc-D-Cys(Bzl)-OH

Formula: C₁₅H₂₁NO₄S
MW: 311.396 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038253
CAS Number: 102830-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR