Boc-D-Glu-OMe

Synonyms: (R)-4-((tert-Butoxycarbonyl)amino)-5-methoxy-5-oxopentanoic acid

4026435.0001
A-3880.0001BAEA 234.53 USD
A-3880.0001BA
Boc-D-Glu-OMe
Boc-D-Glu-OMe

Sequence: Boc-D-Glu-OMe


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₁H₁₉NO₆
MW: 261.274 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00076930
CAS Number: 55227-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR