BACEA-3840

4015252.0005 4015252.0001
A-3840.0001BAEA 222.89 USD
A-3840.0001BA A-3840.0005BA
BACEA-3840
Sequence: Boc-β-cyclohexyl-D-Ala-OH

Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Cha corresponds to hydrogenated phenylalanine, the aromatic nucleus is replaced by a more flexible aliphatic cycle.

Synonym(s): Boc-D-Cha-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR