1-Boc-piperidine-4-carboxylic acid

Synonyms: 1-(tert-Butoxycarbonyl)piperidine-4-carboxylic acidN-BOC-4-Piperidinecarboxylic acidBoc-Inp-OH1-Boc-isonipecotic acid

4026097.0005
A-3815.0005BAEA 149.8 USD
A-3815.0005BA
1-Boc-piperidine-4-carboxylic acid
1-Boc-piperidine-4-carboxylic acid

Sequence: Boc-isonipecotic acid


Nip and Inp derivatives.


Synonym(s): Boc-piperidine-4-carboxylic acid#Boc-Inp-OH

Formula: C₁₁H₁₉NO₄
MW: 229.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00076999
CAS Number: 84358-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR