(S)-N-Boc-4-bromophenylalanine

Synonyms: Boc-p-bromo-Phe-OHBoc-Phe(4-Br)-OH

4025302.0005
A-3695.0005BAEA 757.83 USD
A-3695.0005BA
(S)-N-Boc-4-bromophenylalanine
(S)-N-Boc-4-bromophenylalanine

Sequence: Boc-p-bromo-Phe-OH


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₁₄H₁₈BrNO₄
MW: 344.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00237571
CAS Number: 62129-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR