Boc-S-4-methylbenzyl-D-cysteine

Synonyms: Boc-D-Cys(pMeBzl)-OHBoc-D-Cys(Mbzl)-OH

4019735.0005
A-3545.0005BAEA 615.92 USD
A-3545.0005BA
Boc-S-4-methylbenzyl-D-cysteine
Boc-S-4-methylbenzyl-D-cysteine

Sequence: Boc-D-Cys(Mbzl)-OH

Formula: C₁₆H₂₃NO₄S
MW: 325.423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076924
CAS Number: 61925-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR