Boc-Dab(Boc)-OH·DCHA

Synonyms: Boc-Dab(Boc)-OH·DCHABoc-Dab(Boc)-OH.DCHA(R)-2,3-Di-(Boc-amino)propionic acid dicyclohexylammonium salt

4018915.0001 4018915.0005
A-3480.0005BAEA 633.81 USD
A-3480.0005BA A-3480.0001BA
Boc-Dab(Boc)-OH·DCHA
Boc-Dab(Boc)-OH·DCHA

Sequence: Boc-Dab(Boc)-OH · DCHA

Formula: C₂₆H₄₉N₃O₆
MW: 499.693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238312
CAS Number: 201472-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR