Boc-HCI-OH

Synonyms: Boc-Homocit-OH

4018228.0001
A-3465.0001BAEA 206.68 USD
A-3465.0001BA
Boc-HCI-OH
Boc-HCI-OH

Sequence: Boc-Homocit-OH


Hci derivatives.


Synonym(s): Boc-Lys(carbamoyl)-OH

Formula: C₁₂H₂₃N₃O₅
MW: 289.332 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00151903
CAS Number: 201418-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR