N-Boc-S-methyl-L-cysteine

Synonyms: Boc-Cys(Me)-OH

4018560.0005
A-3455.0005BAEA 289.36 USD
A-3455.0005BA
N-Boc-S-methyl-L-cysteine
N-Boc-S-methyl-L-cysteine

Sequence: Boc-Cys(Me)-OH


Educt for the synthesis of potential inhibitors of the glycosphingolipid biosynthesis (Brenner-Weiss et al.). Cys(Me) (Smc) can be incorporated in peptides as precursor of dehydroalanine. Dha-containing peptides are key intermediates in the synthesis of lantibiotics.

Formula: C₉H₁₇NO₄S
MW: 235.298 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076923
CAS Number: 16947-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR