Boc-N-Me-Arg(Mtr)-OH

4018212.0001 4018212.0005
A-3430.0001BAEA 220.73 USD
A-3430.0001BA A-3430.0005BA
Boc-N-Me-Arg(Mtr)-OH
Boc-N-Me-Arg(Mtr)-OH

Sequence: Boc-N-Me-Arg(Mtr)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also E-2770, H-Arg(Me)-OH · HCl, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₂₂H₃₆N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD00236863
CAS Number: 125602-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR