Boc-D-Orn(fmoc)-OH
Synonyms:
(R)-5-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)pentanoic acid
4019007.0001
4019007.0005
A-3375.0001BAEA
289.36
USD
A-3375.0001BA
A-3375.0005BA
Boc-D-Orn(fmoc)-OH
Boc-D-Orn(fmoc)-OH
Sequence: Boc-D-Orn(Fmoc)-OH
Formula:
C₂₅H₃₀N₂O₆ MW: 454.523 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00151883 CAS Number: 163336-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...