Boc-ASP(OSU)-OBZL

4015685.0005
A-3350.0005BAEA 344.37 USD
A-3350.0005BA
Boc-ASP(OSU)-OBZL
Boc-ASP(OSU)-OBZL

Sequence: Boc-Asp(OSu)-OBzl


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₂₀H₂₄N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD00236828
CAS Number: 140171-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR