BACEA-3340

4019207.1000 4019207.0250
A-3340.1000BAEA 2143.39 USD
A-3340.1000BA A-3340.0250BA
BACEA-3340
Sequence: Boc-cyclohexylstatine
Synonym(s): (3S,4S)-4-(Boc-amino)-5-cyclohexyl-3-hydroxy-pentanoic acid#Boc-ACHPA
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR