Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-Bpa-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-p-Bz-Phe-OH

A-3295.0001 A-3295.0005
A-3295.0001BAEA 203.16 USD InStock
A-3295.0001BA A-3295.0005BA
Boc-Bpa-OH
Boc-Bpa-OH
Sequence: Boc-p-Bz-Phe-OH

Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.
4-Bpa is incorporated into peptides as a photolabel or for photocrosslinking.

Synonym(s): Boc-Bpa-OH


Formula: C₂₁H₂₃NO₅
MW: 369.417 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151886
CAS Number: 104504-43-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty