Boc-Lys(nicotinoyl)-OH

4016403.0005 4016403.0001
A-3135.0005BAEA 509.83 USD
A-3135.0005BA A-3135.0001BA
Boc-Lys(nicotinoyl)-OH
Boc-Lys(nicotinoyl)-OH

Sequence: Boc-Lys(nicotinoyl)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₇H₂₅N₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00076969
CAS Number: 14609-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR