Boc-ARG(PMC)-OH

4014329.0005 4014329.0001
A-2875.0001BAEA 110.22 USD
A-2875.0001BA A-2875.0005BA
Boc-ARG(PMC)-OH
Boc-ARG(PMC)-OH

Sequence: Boc-Arg(Pmc)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₂₅H₄₀N₄O₇S
Storage Temperature: Freezer
MDL Number: MFCD00236823
CAS Number: 200125-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR