BACEA-2830

4014372.0005 4014372.0025 4014372.0001
A-2830.0005BAEA 1239.48 USD
A-2830.0005BA A-2830.0001BA A-2830.0025BA
BACEA-2830
Sequence: Boc-Homopro-OH

Pip (or Hpr) derivatives.

Synonym(s): Boc-L-pipecolic acid#Boc-Pip-OH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR