Boc-ORN(Boc)-OH

Synonyms: Boc-Orn(Boc)-OH

4007556.0025 4007556.0005
A-2645.0025BAEA 633.81 USD
A-2645.0025BA A-2645.0005BA
Boc-ORN(Boc)-OH
Boc-ORN(Boc)-OH

Sequence: Boc-Orn(Boc)-OH

Formula: C₁₅H₂₈N₂O₆
MW: 332.397 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151882
CAS Number: 57133-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR