Boc-d-Asp(ochex)-OH

Synonyms: N-Boc-D-aspartic acid 4-cyclohexyl ester

4013317.0005
A-2625.0005BAEA 522.23 USD
A-2625.0005BA
Boc-d-Asp(ochex)-OH
Boc-d-Asp(ochex)-OH

Sequence: Boc-D-Asp(OcHex)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₅H₂₅NO₆
MW: 315.366 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060741
CAS Number: 112898-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR