Boc-D-Tyr-OMe

Synonyms: N-Boc-D-tyrosine methyl ester

4002487.0005 4002487.0025
A-2425.0005BAEA 165.33 USD
A-2425.0005BA A-2425.0025BA
Boc-D-Tyr-OMe
Boc-D-Tyr-OMe

Sequence: Boc-D-Tyr-OMe


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₅H₂₁NO₅
Storage Temperature: Freezer
MDL Number: MFCD00057824
CAS Number: 76757-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR