Boc-Tyr-OSu

4001331.0005 4001331.0025
A-2415.0025BAEA 496.04 USD
A-2415.0025BA A-2415.0005BA
Boc-Tyr-OSu
Boc-Tyr-OSu

Sequence: Boc-Tyr-OSu


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₈H₂₂N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00037910
CAS Number: 20866-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR