N(α)-Boc-L-tryptophan

Synonyms: Boc-Trp-OHBoc-Trp-OH 

4000240.0100
A-2360.0100BAEA 147.61 USD
A-2360.0100BA
N(α)-Boc-L-tryptophan
N(α)-Boc-L-tryptophan

Sequence: Boc-Trp-OH

Formula: C₁₆H₂₀N₂O₄
MW: 304.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065595
CAS Number: 13139-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR