Boc-D-Pro-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-D-proline(2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid

4000664.0025 4000664.0005
A-2240.0005BAEA 207.49 USD
A-2240.0005BA A-2240.0025BA
Boc-D-Pro-OH
Boc-D-Pro-OH

Sequence: Boc-D-Pro-OH

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063226
CAS Number: 37784-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR