Boc-L-norvaline

Synonyms: N-Boc-L-norvalineN-(tert-Butoxycarbonyl)-L-norvalineBoc-Nva-OH

4005022.0005 4005022.0025
A-2155.0025BAEA 584.45 USD
A-2155.0025BA A-2155.0005BA
Boc-L-norvaline
Boc-L-norvaline

Sequence: Boc-Nva-OH
Synonym(s): Boc-L-2-aminovaleric acid

Formula: C₁₀H₁₉NO₄
MW: 217.265 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037268
CAS Number: 53308-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR