Boc-Nle-OSu

4002808.0005 4002808.0001
A-2150.0005BAEA 330.59 USD
A-2150.0005BA A-2150.0001BA
Boc-Nle-OSu
Boc-Nle-OSu

Sequence: Boc-Nle-OSu
Synonym(s): Boc-L-2-aminohexanoic acid-OSu

Formula: C₁₅H₂₄N₂O₆
Storage Temperature: Freezer
MDL Number: MFCD00190816
CAS Number: 36360-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR