Boc-N-methyl-L-leucine

Synonyms: Boc-N-Me-Leu-OH

4002308.0001
A-2060.0001BAEA 96.55 USD
A-2060.0001BA
Boc-N-methyl-L-leucine
Boc-N-methyl-L-leucine

Sequence: Boc-N-Me-Leu-OH


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038522
CAS Number: 53363-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR