Boc-Met(O)-Oh

Synonyms: N-Boc-L-methionine sulphoxide

4010049.0005
A-1965.0005BAEA 96.55 USD
A-1965.0005BA
Boc-Met(O)-Oh
Boc-Met(O)-Oh

Sequence: Boc-Met(O)-OH


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.


(Diastereomeric mixture) Met(O) can be reduced to Met in peptides containing disulfide bridges with ammonium iodide in TFA in the presence of dimethyl sulfide.

Formula: C₁₀H₁₉NO₅S
Storage Temperature: Ambient
MDL Number: MFCD00037172
CAS Number: 34805-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR