Boc-His-OMe

Supplier: BACHEM AMERICAS INC

Synonyms: N-Boc-L-histidine methyl ester

4000662.0005 4000662.0001
A-1775.0005BAEA 298.4 USD InStock
A-1775.0005BA A-1775.0001BA
Boc-His-OMe
Boc-His-OMe

Sequence: Boc-His-OMe


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₁₂H₁₉N₃O₄
MW: 269,301 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00167635
CAS Number: 2488-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR