N(α)-Boc-D-histidine
Synonyms: Boc-D-His-OH
4007022.0005
A-1770.0005BAEA 292.23 USD
A-1770.0005BA
N(α)-Boc-D-histidine
N(α)-Boc-D-histidine
Sequence: Boc-D-His-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.