Boc-D-Glu(OtBu)-OH

Synonyms: N-Boc-D-glutamic acid 5-tert-butyl ester

4005552.0005
A-1680.0005BAEA 137.74 USD
A-1680.0005BA
Boc-D-Glu(OtBu)-OH
Boc-D-Glu(OtBu)-OH

Sequence: Boc-D-Glu(OtBu)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₁₄H₂₅NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD00076927
CAS Number: 104719-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR