Boc-Lys(fmoc)-OMe

Synonyms: (S)-Methyl 6-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)hexanoateNα-Boc-Nepsilon-Fmoc-L-lysine methyl ester

4004102.0001
A-1615.0001BAEA 220.73 USD
A-1615.0001BA
Boc-Lys(fmoc)-OMe
Boc-Lys(fmoc)-OMe

Sequence: Boc-Lys(Fmoc)-OMe


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₇H₃₄N₂O₆
MW: 482.577 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236856
CAS Number: 133628-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR