Boc-D-Lys(Z)-OH

Synonyms: Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine

4003809.0005
A-1470.0005BAEA 234.53 USD
A-1470.0005BA
Boc-D-Lys(Z)-OH
Boc-D-Lys(Z)-OH

Sequence: Boc-D-Lys(Z)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₁₉H₂₈N₂O₆
MW: 380.441 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038262
CAS Number: 55878-47-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR