Boc-His(Z)-OH

Supplier: BACHEM AMERICAS INC

4009724.0025
A-1460.0025BAEA 762 USD
A-1460.0025BA
Boc-His(Z)-OH
Boc-His(Z)-OH

Sequence: Boc-His(Z)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₁₉H₂₃N₃O₆
Storage Temperature: Freezer
MDL Number: MFCD00076942
CAS Number: 50305-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR