Boc-D-Thr(tBu)-OH

Synonyms: (2R,3S)-3-(tert-Butoxy)-2-((tert-butoxycarbonyl)amino)butanoic acid

4007520.0005 4007520.0001
A-1445.0005BAEA 672.09 USD
A-1445.0005BA A-1445.0001BA
Boc-D-Thr(tBu)-OH
Boc-D-Thr(tBu)-OH

Sequence: Boc-D-Thr(tBu)-OH


Bachem also offers phosphothreonine derivatives.

Formula: C₁₃H₂₅NO₅
MW: 275.345 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236876
CAS Number: 201217-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR