Boc-D-Ser(tBu)-OH·DCHA

Synonyms: Boc-D-Ser(tBu)-OH·DCHABoc-D-Ser(tBu)-OH.DCHA

4004211.0001
A-1435.0001BAEA 165.33 USD
A-1435.0001BA
Boc-D-Ser(tBu)-OH·DCHA
Boc-D-Ser(tBu)-OH·DCHA

Sequence: Boc-D-Ser(tBu)-OH · DCHA


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₂₄H₄₆N₂O₅
MW: 442.641 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00236872
CAS Number: 248921-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR