Boc-Tyr(2-Br-Z)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Tyr(2-bromo-Z)-OH

4001250.0100
A-1420.0100BAEA 577.41 USD
A-1420.0100BA
Boc-Tyr(2-Br-Z)-OH
Boc-Tyr(2-Br-Z)-OH

Sequence: Boc-Tyr(2-bromo-Z)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.


Synonym(s): Boc-Tyr(2-Br-Z)-OH

Formula: C₂₂H₂₄BrNO₇
MW: 494.338 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037098
CAS Number: 47689-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR