Boc-D-Ser(BZL)-OSu

4007051.0005
A-1345.0005BAEA 442.3 USD
A-1345.0005BA
Boc-D-Ser(BZL)-OSu
Boc-D-Ser(BZL)-OSu

Sequence: Boc-D-Ser(Bzl)-OSu


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₁₉H₂₄N₂O₇
Storage Temperature: Freezer
MDL Number: MFCD00236873
CAS Number: 82155-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR