Boc-Asp(OtBu)-ONp

4003775.0001
A-1280.0001BAEA 96.55 USD
A-1280.0001BA
Boc-Asp(OtBu)-ONp
Boc-Asp(OtBu)-ONp

Sequence: Boc-Asp(OtBu)-ONp


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.

Formula: C₁₉H₂₆N₂O₈
Storage Temperature: Freezer
MDL Number: MFCD00076913
CAS Number: 29365-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR