Boc-D-Asp(OtBu)-OH·DCHA

Synonyms: Boc-D-Asp(OtBu)-OH.DCHABoc-D-Asp(OtBu)-OH·DCHA

4005551.0005 4005551.0001
A-1270.0001BAEA 124.02 USD
A-1270.0001BA A-1270.0005BA
Boc-D-Asp(OtBu)-OH·DCHA
Boc-D-Asp(OtBu)-OH·DCHA

Sequence: Boc-D-Asp(OtBu)-OH · DCHA


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


CAS-No. (free acid): 155542-33-9 .

Formula: C₂₅H₄₆N₂O₆
MW: 470.651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00076911
CAS Number: 200334-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR