AC1L70HP

Synonyms: Boc-Asn-OSu

4006410.0005 4006410.0025
A-1225.0005BAEA 234.53 USD
A-1225.0005BA A-1225.0025BA
AC1L70HP
AC1L70HP

Sequence: Boc-Asn-OSu


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₁₃H₁₉N₃O₇
MW: 329.309 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00190785
CAS Number: 42002-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR