(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-(Boc-amino)-4-phenylbutyric AcidN-Boc-L-homophenylalanineBoc-Homophe-OH(S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid

4009210.0005 4009210.0001
A-1190.0005BAEA 321.85 USD
A-1190.0005BA A-1190.0001BA
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Sequence: Boc-Homophe-OH


Hph derivatives. Bachem also offers Fmoc-Hph-Wang resin, D-2005.


Synonym(s): (S)-2-(Boc-amino)-4-phenyl-butyric acid

Formula: C₁₅H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD00076904
CAS Number: 100564-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR