Boc-Abu-OH

Synonyms: Boc-L-2-aminobutanoic acidBOC-L-α-Aminobutyric acid(S)-2-(tert-Butoxycarbonylamino)butyric acidBoc-L-Abu-OH(S)-2-(Boc-amino)butyric acid

4002314.0005 4002314.0025
A-1135.0005BAEA 192.89 USD
A-1135.0005BA A-1135.0025BA
Boc-Abu-OH
Boc-Abu-OH

Sequence: Boc-Abu-OH

Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037267
CAS Number: 34306-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR