Boc-b-Alanine

Synonyms: N-Boc-beta-alanineBoc-β-Ala-OHBOC-beta-alanine

4001867.0100 4001867.0025
A-1075.0025BAEA 101.37 USD
A-1075.0025BA A-1075.0100BA
Boc-b-Alanine
Boc-b-Alanine

Sequence: Boc-β-Ala-OH


Bachem also offers Fmoc-β-Ala-Wang resin (D-1625) and Boc-β-Ala-PAM resin (D-1650).

Formula: C₈H₁₅NO₄
MW: 189.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037291
CAS Number: 3303-84-2
EINECS: 221-979-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR