Boc-Tyr(AC)-OH

4010204.0005 4010204.0001
A-1055.0005BAEA 385.8 USD
A-1055.0005BA A-1055.0001BA
Boc-Tyr(AC)-OH
Boc-Tyr(AC)-OH

Sequence: Boc-Tyr(Ac)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₆H₂₁NO₆
Storage Temperature: Ambient
MDL Number: MFCD00057973
CAS Number: 80971-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR