Boc-Lys(boc)-ONP

Synonyms: N2,N6-Di-Boc-L-Lysine 4-Nitrophenyl EsterNα,Nepsilon-Di-Boc-L-lysine 4-nitrophenyl ester

4009543.0005 4009543.0025
A-1030.0005BAEA 96.52 USD
A-1030.0005BA A-1030.0025BA
Boc-Lys(boc)-ONP
Boc-Lys(boc)-ONP

Sequence: Boc-Lys(Boc)-ONp


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₂H₃₃N₃O₈
MW: 467.519 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00076961
CAS Number: 2592-19-0
EINECS: 219-988-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR